siRNA Details
| Gene Symbol: HUNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014586.1 | NP_055401.1 | 2 | 668 | ||
| XM_011529537.2 | XP_011527839.1 | 2 | 332 | |||
| GenBank mRNA | AJ271722.1 | - | 2 | 668 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HUNK| Gene Symbol: HUNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014586.1 | NP_055401.1 | 2 | 668 | ||
| XM_011529537.2 | XP_011527839.1 | 2 | 332 | |||
| GenBank mRNA | AJ271722.1 | - | 2 | 668 | ||