siRNA Details
| Gene Symbol: Styk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172891.2 | NP_766479.1 | 9 | 2651 | ||
| GenBank mRNA | AK033606.1 | - | 9 | 2651 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Styk1| Gene Symbol: Styk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172891.2 | NP_766479.1 | 9 | 2651 | ||
| GenBank mRNA | AK033606.1 | - | 9 | 2651 | ||