siRNA Details
| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172898.3 | NP_766486.1 | 15 | 2895 | ||
| GenBank mRNA | AK049284.1 | - | 15 | 2896 | ||
| AK083069.1 | - | 13 | 3445 | |||
| BC052773.1 | - | 15 | 2358 | 6314924 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kirrel2| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172898.3 | NP_766486.1 | 15 | 2895 | ||
| GenBank mRNA | AK049284.1 | - | 15 | 2896 | ||
| AK083069.1 | - | 13 | 3445 | |||
| BC052773.1 | - | 15 | 2358 | 6314924 | ||