siRNA Details
| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172926.3 | NP_766514.2 | 1, 2 | 292 | ||
| GenBank mRNA | AK046839.1 | - | 1, 2 | 234 | ||
| AK049371.1 | - | 1, 2 | 271 | |||
| AK082389.1 | - | 1, 2 | 271 | |||
| BC080799.1 | - | 1, 2 | 277 | 4948538 | ||
| BY013426.1 | - | 1, 2 | 294 | |||

Snx14
Genomic Map