siRNA Details
| Gene Symbol: Ankrd13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172945.2 | NP_766533.2 | 16 | 2373 | ||
| GenBank mRNA | AK081149.1 | - | 1310 | |||
| BC022693.1 | - | 5 | 919 | |||
| BC039786.1 | - | 5 | 1337 | |||
| BC056928.1 | - | 9 | 1859 | |||
| BC071189.1 | - | 10 | 1459 | |||
| BC098207.1 | - | 7 | 1669 | |||
| BC151158.1 | - | 15 | 2130 | 9088046 | ||
| BC172028.1 | - | 15 | 2131 | |||

Ankrd13b
Genomic Map