siRNA Details
| Gene Symbol: Sntg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172951.3 | NP_766539.2 | 2 | 304 | ||
| GenBank mRNA | AF367760.1 | - | 2 | 159 | ||
| AK008673.1 | - | 2 | 235 | |||
| AK035310.1 | - | 2 | 258 | |||
| AK087560.1 | - | 2 | 259 | |||
| AK137345.1 | - | 2 | 263 | |||
| BC058204.1 | - | 2 | 194 | |||
| BC138956.1 | - | 2 | 218 | 8861978 | ||
| BC145476.1 | - | 2 | 218 | |||
| BY098098.1 | - | 2 | 305 | |||

Sntg2
Genomic Map