siRNA Details
| Gene Symbol: 4930503L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172967.2 | NP_766555.1 | 3 | 710 | ||
| GenBank mRNA | AK037066.1 | - | 3 | 711 | ||
| AK080780.1 | - | 3 | 741 | |||
| AK087788.1 | - | 3 | 683 | |||
| AK148410.1 | - | 3 | 734 | |||
| AK160248.1 | - | 3 | 584 | |||
| AK160714.1 | - | 3 | 680 | |||
| BC049726.1 | - | 222 | ||||
| BC057927.1 | - | 2 | 427 | 5290837 | ||
| HQ438967.1 | - | 2 | 222 | |||
| HQ438968.1 | - | 2 | 222 | |||
| HQ438969.1 | - | 2 | 222 | |||
| HQ438970.1 | - | 2 | 222 | |||
| HQ438971.1 | - | 2 | 222 | |||
| HQ438972.1 | - | 2 | 222 | |||
| HQ438973.1 | - | 2 | 231 | |||
| HQ438974.1 | - | 2 | 231 | |||
| HQ438975.1 | - | 2 | 231 | |||
| HQ438976.1 | - | 1 | 181 | |||
| HQ438977.1 | - | 1 | 181 | |||
| HQ438978.1 | - | 1 | 190 | |||
| HQ438979.1 | - | 1 | 181 | |||
| HQ438980.1 | - | 1 | 181 | |||
| HQ438981.1 | - | 1 | 181 | |||
| HQ438982.1 | - | 1 | 181 | |||
| HQ438983.1 | - | 1 | 181 | |||
| HQ438984.1 | - | 1 | 190 | |||
| HQ438985.1 | - | 1 | 181 | |||
| HQ438986.1 | - | 1 | 190 | |||
| HQ438987.1 | - | 1 | 181 | |||
| HQ438988.1 | - | 1 | 181 | |||
| HQ438989.1 | - | 1 | 181 | |||
| HQ438990.1 | - | 1 | 190 | |||
| HQ438991.1 | - | 1 | 190 | |||
| HQ438992.1 | - | 1 | 181 | |||
| HQ438993.1 | - | 1 | 190 | |||
| HQ438994.1 | - | 1 | 181 | |||
| HQ438995.1 | - | 1 | 190 | |||
| HQ438996.1 | - | 1 | 190 | |||
| HQ438997.1 | - | 1 | 190 | |||
| HQ438998.1 | - | 2 | 231 | |||
| HQ438999.1 | - | 1 | 181 | |||
| HQ439000.1 | - | 1 | 181 | |||
| HQ439001.1 | - | 1 | 190 | |||
| HQ439002.1 | - | 1 | 181 | |||
| HQ439003.1 | - | 1 | 181 | |||
| HQ439004.1 | - | 1 | 190 | |||
| HQ439005.1 | - | 1 | 181 | |||
| HQ439006.1 | - | 1 | 181 | |||
| HQ439007.1 | - | 1 | 190 | |||
| HQ439008.1 | - | 1 | 190 | |||
| HQ439009.1 | - | 1 | 181 | |||
| HQ439010.1 | - | 1 | 190 | |||
| HQ439011.1 | - | 1 | 190 | |||
| HQ439012.1 | - | 1 | 181 | |||

4930503L19Rik
Genomic Map