siRNA Details
| Gene Symbol: Gtf3c4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172977.3 | NP_766565.2 | 1, 2 | 486 | ||
| GenBank mRNA | AK030351.1 | - | 1, 2 | 413 | ||
| AK141269.1 | - | 1, 2 | 467 | |||
| BC061476.1 | - | 1, 2 | 419 | |||
| BC095998.1 | - | 1, 2 | 425 | 30736671 | ||
| BC117919.1 | - | 1, 2 | 398 | 40097451 | ||
| BC117920.1 | - | 1, 2 | 396 | 40097456 | ||
| CN530016.1 | - | 1, 2 | 486 | |||

Gtf3c4
Genomic Map