siRNA Details
| Gene Symbol: Nkain3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290410.1 | NP_001277339.1 | 2 | 432 | ||
| NM_172987.2 | NP_766575.2 | 2 | 432 | |||
| GenBank mRNA | AK049776.1 | - | 2 | 632 | ||
| AK053823.1 | - | 2 | 317 | |||
| AK141456.1 | - | 2 | 332 | |||
| BC075683.1 | - | 2 | 432 | 30613986 | ||
| EF058052.1 | - | 2 | 79 | |||
| EF058053.1 | - | 2 | 79 | |||
| EF058054.1 | - | 2 | 79 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109540, XM_008763597.

Nkain3
Genomic Map