siRNA Details
| Gene Symbol: Fbxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172988.4 | NP_766576.1 | 3 | 398 | ||
| GenBank mRNA | AK028786.1 | - | 3 | 382 | ||
| AK032391.1 | - | 3 | 377 | |||
| AK051429.1 | - | 3 | 349 | |||
| AK083661.1 | - | 3 | 372 | |||
| BC033265.1 | - | 3 | 325 | 3983942 | ||
| BY128470.1 | - | 3 | 262 | |||

Fbxl4
Genomic Map