siRNA Details
Gene Symbol: Ip6k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173027.2 | NP_766615.1 | 2 | 349 | ||
GenBank mRNA | AK052857.1 | - | 2 | 350 | ||
BC141263.1 | - | 2 | 243 | 9055875 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ip6k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173027.2 | NP_766615.1 | 2 | 349 | ||
GenBank mRNA | AK052857.1 | - | 2 | 350 | ||
BC141263.1 | - | 2 | 243 | 9055875 |