siRNA Details
| Gene Symbol: AFG3L1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003226.1 | 10 | 1144 | |||
| NR_003227.1 | 9 | 971 | ||||
| NR_003228.1 | 10 | 1110 | ||||
| GenBank mRNA | AF329691.1 | - | 9 | 895 | ||
| AF329692.1 | - | 4 | 446 | |||
| AJ001495.1 | - | 2 | 118 | |||
| AK056488.1 | - | 10 | 1115 | |||
| AK091293.1 | - | 10 | 1107 | |||
| AK097044.1 | - | 9 | 958 | |||
| AK126911.1 | - | 8 | 935 | |||
| AK128352.1 | - | 3 | 810 | |||
| AK128804.1 | - | 4 | 337 | |||
| AL832984.1 | - | 4 | 368 | |||
| BC021245.2 | - | 8 | 717 | |||

AFG3L1P
Genomic Map