siRNA Details
| Gene Symbol: AI118078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172923.3 | NP_766511.1 | 7, 8 | 797 | ||
| GenBank mRNA | AK036040.1 | - | 7, 8 | 797 | ||
| BC068128.1 | - | 8, 9 | 953 | 6835202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI118078| Gene Symbol: AI118078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172923.3 | NP_766511.1 | 7, 8 | 797 | ||
| GenBank mRNA | AK036040.1 | - | 7, 8 | 797 | ||
| BC068128.1 | - | 8, 9 | 953 | 6835202 | ||