siRNA Details
| Gene Symbol: Dclk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172928.5 | NP_766516.2 | 2 | 874 | ||
| GenBank mRNA | AK050312.1 | - | 2 | 842 | ||
| AK135010.1 | - | 2 | 863 | |||
| BC056929.1 | - | 1 | 542 | 6816292 | ||
| DQ286388.1 | - | 2 | 843 | |||

Dclk3
Genomic Map