siRNA Details
| Gene Symbol: Gm14085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085518.1 | NP_001078987.1 | 3, 4 | 1350 | ||
| Gene Symbol: Slc28a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172980.2 | NP_766568.1 | 4, 5 | 1512 | ||
| GenBank mRNA | AB485921.1 | - | 4, 5 | 811 | ||
| AF079853.2 | - | 3, 4 | 387 | |||
| AK080957.1 | - | 4, 5 | 1513 | |||
| AK152776.1 | - | 3, 4 | 447 | |||

Gm14085
Genomic Map