siRNA Details
| Gene Symbol: Nkain3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290410.1 | NP_001277339.1 | 3, 4 | 634 | ||
| NM_172987.2 | NP_766575.2 | 3, 4 | 634 | |||
| GenBank mRNA | AK049776.1 | - | 3, 4 | 834 | ||
| AK053823.1 | - | 3, 4 | 519 | |||
| AK141456.1 | - | 3, 4 | 534 | |||
| BC075683.1 | - | 3, 4 | 634 | 30613986 | ||
| EF058052.1 | - | 3, 4 | 281 | |||
| EF058053.1 | - | 3, 4 | 281 | |||
| EF058054.1 | - | 3, 4 | 281 | |||

Nkain3
Genomic Map