siRNA Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173011.2 | NP_766599.2 | 3 | 367 | ||
| GenBank mRNA | AF212319.1 | - | 3 | 338 | ||
| AK088110.1 | - | 3 | 339 | |||
| AK145753.1 | - | 3 | 312 | |||
| AK146564.1 | - | 3 | 350 | |||
| AK161640.1 | - | 3 | 351 | |||
| AK169395.1 | - | 3 | 364 | |||
| AK212860.1 | - | 1 | 72 | |||
| BC060030.1 | - | 3 | 355 | 6515657 | ||

Idh2
Genomic Map