siRNA Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172909.4 | NP_766497.3 | 14 | 3408 | ||
| GenBank mRNA | AF539744.1 | - | 14 | 3164 | ||
| AK046675.1 | - | 13 | 3585 | |||
| AK088010.1 | - | 13 | 3401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd163l1| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172909.4 | NP_766497.3 | 14 | 3408 | ||
| GenBank mRNA | AF539744.1 | - | 14 | 3164 | ||
| AK046675.1 | - | 13 | 3585 | |||
| AK088010.1 | - | 13 | 3401 | |||