siRNA Details
| Gene Symbol: Zkscan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130529.2 | NP_001124001.1 | 10 | 3080 | ||
| NM_001291014.1 | NP_001277943.1 | 10 | 2834 | |||
| NM_172941.4 | NP_766529.3 | 10 | 3127 | |||
| GenBank mRNA | AK051711.1 | - | 10 | 2416 | ||
| AK155196.1 | - | 10 | 2657 | |||
| AK155490.1 | - | 10 | 2709 | |||
| AK157002.1 | - | 4 | 789 | |||
| AK170924.1 | - | 10 | 2657 | |||
| AK170989.1 | - | 10 | 2662 | |||
| AK171190.1 | - | 10 | 2709 | |||
| AY302242.1 | - | 9 | 2608 | |||
| BC040205.1 | - | 9 | 2612 | 4947290 | ||

Zkscan17
Genomic Map