siRNA Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170978.1 | NP_001164449.1 | 15 | 1630 | ||
| NM_172961.3 | NP_766549.2 | 16 | 1798 | |||
| GenBank mRNA | AK036128.1 | - | 15 | 1575 | ||
| AK132282.1 | - | 16 | 1747 | |||
| AK156736.1 | - | 4 | 507 | |||
| AK165348.1 | - | 16 | 1749 | |||
| BC058079.1 | - | 16 | 1738 | 6826741 | ||
| BC058521.1 | - | 16 | 1738 | 6825487 | ||

Abat
Genomic Map