siRNA Details
| Gene Symbol: Mon1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048143.2 | NP_001041608.1 | 6 | 2191 | ||
| NM_173015.3 | NP_766603.1 | 5 | 2381 | |||
| GenBank mRNA | AK030288.1 | - | 5 | 2347 | ||
| AK157390.1 | - | 6 | 2195 | |||
| AK173047.1 | - | 5 | 2053 | |||
| BC062639.1 | - | 5 | 2057 | |||

Mon1b
Genomic Map