siRNA Details
Gene Symbol: BC024139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142968.1 | NP_001136440.1 | 13 | 1987 | ||
GenBank mRNA | AK078032.1 | - | 13 | 1975 | ||
AK081549.1 | - | 13 | 1989 | |||
BC060242.1 | - | 13 | 2030 | |||
BC063071.1 | - | 8 | 1425 | 5694464 |
