siRNA Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161851.2 | NP_001155323.1 | 2, 3 | 372 | ||
| NM_001161852.2 | NP_001155324.1 | 2, 3 | 649 | |||
| NM_173401.5 | NP_775577.2 | 2, 3 | 649 | |||
| GenBank mRNA | AK019407.1 | - | 2, 3 | 404 | ||
| AK077452.1 | - | 2, 3 | 435 | |||
| AK170958.1 | - | 2, 3 | 332 | |||
| AK177996.1 | - | 2, 3 | 367 | |||
| AK178346.1 | - | 2, 3 | 364 | |||
| AK191511.1 | - | 1, 2 | 245 | |||
| AK218805.1 | - | 1, 2 | 178 | |||
| BC028884.1 | - | 2, 3 | 441 | 4165174 | ||
| BM932876.1 | - | 1, 2 | 298 | |||
| BQ042735.1 | - | 2, 3 | 432 | |||
| BY252242.1 | - | 2, 3 | 359 | |||
| CX202860.1 | - | 1, 2 | 73 | |||

Fbxo44
Genomic Map