siRNA Details
Gene Symbol: Jazf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173406.3 | NP_775582.2 | 1, 2 | 167 | ||
GenBank mRNA | AK043767.1 | - | 1, 2 | 117 | ||
AK050474.1 | - | 1, 2 | 139 | |||
BY278789.1 | - | 1, 2 | 167 |
