siRNA Details
| Gene Symbol: Synb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173420.3 | NP_775596.1 | 938 | |||
| GenBank mRNA | AK086309.1 | - | 939 | |||
| BC137938.1 | - | 680 | 8860957 | |||
| BC137956.1 | - | 680 | 8860975 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synb| Gene Symbol: Synb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173420.3 | NP_775596.1 | 938 | |||
| GenBank mRNA | AK086309.1 | - | 939 | |||
| BC137938.1 | - | 680 | 8860957 | |||
| BC137956.1 | - | 680 | 8860975 | |||