siRNA Details
| Gene Symbol: 4930595M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173435.3 | NP_775611.2 | 1 | 233 | ||
| GenBank mRNA | AK029824.1 | - | 1 | 234 | ||
| BB616016.1 | - | 1 | 234 | |||
| BC131910.1 | - | 1 | 229 | 40130187 | ||
| BC132591.1 | - | 1 | 229 | 40130868 | ||

4930595M18Rik
Genomic Map