siRNA Details
| Gene Symbol: Nrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173440.2 | NP_775616.1 | 4 | 798 | ||
| GenBank mRNA | AF053062.1 | - | 2 | 446 | ||
| AK036273.1 | - | 4 | 799 | |||
| AK156233.1 | - | 4 | 777 | |||
| AK157467.1 | - | 4 | 777 | |||
| AK207663.1 | - | 1 | 116 | |||
| BC060232.1 | - | 3 | 596 | 6852643 | ||
| BC075638.1 | - | 4 | 675 | |||

Nrip1
Genomic Map