siRNA Details
| Gene Symbol: Osbpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134791.2 | NP_001128263.1 | 12 | 1073 | ||
| NM_133885.4 | NP_598646.4 | 15 | 1306 | |||
| NM_173350.3 | NP_775485.2 | 13 | 1112 | |||
| GenBank mRNA | AK035658.1 | - | 13 | 1061 | ||
| AK075981.1 | - | 2 | 91 | |||
| AK131984.1 | - | 15 | 1277 | |||
| AK172010.1 | - | 13 | 1099 | |||
| BC021507.1 | - | 11 | 934 | 5347121 | ||
| BC023759.1 | - | 15 | 1261 | |||
| BC026927.1 | - | 13 | 995 | 5134780 | ||
| BC089163.1 | - | 12 | 1014 | 5367603 | ||

Osbpl9
Genomic Map