siRNA Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173364.5 | NP_775540.4 | 7 | 2107 | ||
| GenBank mRNA | AK036132.1 | - | 1 | 270 | ||
| AY341877.1 | - | 6 | 1948 | |||
| BC027167.1 | - | 1 | 853 | |||
| BC034572.1 | - | 1 | 717 | |||
| BC055684.1 | - | 1 | 404 | |||
| BC099896.1 | - | 6 | 1919 | 30545532 | ||
| BC099930.1 | - | 7 | 2052 | 30058096 | ||

Zfp445
Genomic Map