siRNA Details
| Gene Symbol: Proser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173382.1 | NP_775558.2 | 3 | 626 | ||
| GenBank mRNA | AK034174.1 | - | 3 | 777 | ||
| AK038404.1 | - | 3 | 626 | |||
| AK087909.1 | - | 3 | 647 | |||
| AK161440.1 | - | 2 | 445 | |||
| AK199697.1 | - | 3 | 191 | |||
| AK207125.1 | - | 3 | 191 | |||
| BC086674.1 | - | 3 | 183 | 30653602 | ||

Proser1
Genomic Map