siRNA Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173392.4 | NP_775568.1 | 3 | 394 | ||
| GenBank mRNA | AK045090.1 | - | 3 | 452 | ||
| AK122241.1 | - | 3 | 375 | |||
| AK170435.1 | - | 3 | 413 | |||
| BC042669.1 | - | 3 | 392 | 4235024 | ||

Zfyve16
Genomic Map