siRNA Details
| Gene Symbol: BC030500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173411.2 | NP_775587.1 | 3 | 252 | ||
| GenBank mRNA | BC030500.1 | - | 3 | 263 | 5401612 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC030500| Gene Symbol: BC030500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173411.2 | NP_775587.1 | 3 | 252 | ||
| GenBank mRNA | BC030500.1 | - | 3 | 263 | 5401612 | |