siRNA Details
| Gene Symbol: Nyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173415.4 | NP_775591.1 | 3 | 1701 | ||
| GenBank mRNA | AK038912.1 | - | 1 | 1095 | ||
| AY114303.1 | - | 3401 | ||||
| BC035244.1 | - | 3 | 1701 | 5362608 | ||
| BC048381.1 | - | 3 | 1700 | 5362608 | ||

Nyx
Genomic Map