siRNA Details
| Gene Symbol: Fam124b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173425.3 | NP_775601.1 | 1 | 434 | ||
| GenBank mRNA | AK043867.1 | - | 1 | 435 | ||
| BC150872.1 | - | 1 | 319 | 9087783 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam124b| Gene Symbol: Fam124b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173425.3 | NP_775601.1 | 1 | 434 | ||
| GenBank mRNA | AK043867.1 | - | 1 | 435 | ||
| BC150872.1 | - | 1 | 319 | 9087783 | ||