siRNA Details
| Gene Symbol: Fam110b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173426.2 | NP_775602.1 | 3 | 816 | ||
| GenBank mRNA | AK052596.1 | - | 3 | 817 | ||
| AK077232.1 | - | 1 | 448 | |||
| AK146011.1 | - | 2 | 574 | |||
| AK159054.1 | - | 3 | 825 | |||
| BC055683.1 | - | 3 | 746 | 6414160 | ||

Fam110b
Genomic Map