siRNA Details
| Gene Symbol: Fkrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173430.2 | NP_775606.1 | 3 | 1668 | ||
| GenBank mRNA | AJ511806.1 | - | 3 | 1651 | ||
| AK080624.1 | - | 1 | 1478 | |||
| AK135089.1 | - | 3 | 1668 | |||
| AK156156.1 | - | 3 | 1667 | |||
| AK166068.1 | - | 3 | 1670 | |||
| AK168308.1 | - | 3 | 1666 | |||
| BC053072.1 | - | 3 | 1651 | 5686286 | ||

Fkrp
Genomic Map