siRNA Details
| Gene Symbol: Ilvbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173751.4 | NP_776112.1 | 3 | 478 | ||
| GenBank mRNA | AK087956.1 | - | 2 | 352 | ||
| AK133553.1 | - | 2 | 408 | |||
| AK138031.1 | - | 2 | 327 | |||
| AK143918.1 | - | 2 | 377 | |||
| AK146075.1 | - | 2 | 406 | |||
| AK169899.1 | - | 2 | 404 | |||
| BC052054.1 | - | 2 | 812 | |||
| BC056459.1 | - | 3 | 436 | 5718157 | ||
| CD768274.1 | - | 3 | 482 | |||

Ilvbl
Genomic Map