siRNA Details
| Gene Symbol: Fnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173753.4 | NP_776114.2 | 16 | 3351 | ||
| GenBank mRNA | AK042779.1 | - | 3 | 816 | ||
| AK047814.1 | - | 3 | 592 | |||
| AK122574.1 | - | 16 | 3299 | |||
| BC024776.1 | - | 15 | ||||
| BC079892.1 | - | 16 | 3341 | 30543296 | ||

Fnip1
Genomic Map