siRNA Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173364.5 | NP_775540.4 | 7 | 3972 | ||
| GenBank mRNA | AK032835.1 | - | 1 | 167 | ||
| AK035697.1 | - | 1 | 328 | |||
| AK036132.1 | - | 1 | 2135 | |||
| BC027167.1 | - | 1 | 2718 | |||
| BC055684.1 | - | 1 | 2269 | |||
| BC099896.1 | - | 6 | 3859 | 30545532 | ||
| BC099930.1 | - | 7 | 3917 | 30058096 | ||

Zfp445
Genomic Map