siRNA Details
| Gene Symbol: Trp53bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173378.2 | NP_775554.2 | 18 | 3723 | ||
| GenBank mRNA | AK133930.1 | - | 3 | 361 | ||
| BC025187.1 | - | 3 | 368 | |||
| BC030894.1 | - | 10 | 2352 | |||
| BC042874.1 | - | 17 | 3325 | |||

Trp53bp2
Genomic Map