siRNA Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173392.4 | NP_775568.1 | 13 | 3927 | ||
| GenBank mRNA | AK080779.1 | - | 9 | 1736 | ||
| AK122241.1 | - | 13 | 3908 | |||
| BC042669.1 | - | 13 | 3925 | 4235024 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfyve16| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173392.4 | NP_775568.1 | 13 | 3927 | ||
| GenBank mRNA | AK080779.1 | - | 9 | 1736 | ||
| AK122241.1 | - | 13 | 3908 | |||
| BC042669.1 | - | 13 | 3925 | 4235024 | ||