siRNA Details
Gene Symbol: Gpr171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173398.3 | NP_775574.1 | 2 | 1879 | ||
GenBank mRNA | AK089287.1 | - | 2 | 1832 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
