siRNA Details
| Gene Symbol: Haus6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173400.2 | NP_775576.2 | 9 | 1248 | ||
| GenBank mRNA | AK051016.1 | - | 9 | 1269 | ||
| AK077681.1 | - | 9 | 1270 | |||
| AK169222.1 | - | 9 | 1250 | |||
| AK173206.1 | - | 9 | 1220 | |||
| BC064725.1 | - | 4 | 445 | |||
| BC068237.1 | - | 9 | 1248 | 30550558 | ||

Haus6
Genomic Map