siRNA Details
| Gene Symbol: Lancl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173414.3 | NP_775590.2 | 5 | 3213 | ||
| GenBank mRNA | AK031623.1 | - | 5 | 3214 | ||
| AK043841.1 | - | 5 | 2414 | |||
| AK045254.1 | - | 5 | 2801 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lancl3| Gene Symbol: Lancl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173414.3 | NP_775590.2 | 5 | 3213 | ||
| GenBank mRNA | AK031623.1 | - | 5 | 3214 | ||
| AK043841.1 | - | 5 | 2414 | |||
| AK045254.1 | - | 5 | 2801 | |||