siRNA Details
| Gene Symbol: Erich5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173421.2 | NP_775597.1 | 3 | 1748 | ||
| GenBank mRNA | BC030476.1 | - | 3 | 1756 | 5368170 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich5| Gene Symbol: Erich5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173421.2 | NP_775597.1 | 3 | 1748 | ||
| GenBank mRNA | BC030476.1 | - | 3 | 1756 | 5368170 | |