siRNA Details
| Gene Symbol: Fem1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173423.4 | NP_775599.1 | 3 | 2642 | ||
| GenBank mRNA | AK028330.1 | - | 3 | 2642 | ||
| AK129440.1 | - | 2575 | ||||
| AK147729.1 | - | 3 | 2532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fem1c| Gene Symbol: Fem1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173423.4 | NP_775599.1 | 3 | 2642 | ||
| GenBank mRNA | AK028330.1 | - | 3 | 2642 | ||
| AK129440.1 | - | 2575 | ||||
| AK147729.1 | - | 3 | 2532 | |||