siRNA Details
| Gene Symbol: Pskh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173432.2 | NP_775608.1 | 3 | 1707 | ||
| GenBank mRNA | AK053397.1 | - | 3 | 1741 | ||
| AK153861.1 | - | 3 | 1754 | |||
| AK154570.1 | - | 3 | 1737 | |||
| AK166338.1 | - | 3 | 1731 | |||
| BC050128.1 | - | 3 | 1707 | 30024345 | ||

Pskh1
Genomic Map