siRNA Details
| Gene Symbol: Arsj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173451.3 | NP_775627.1 | 2 | 1862 | ||
| GenBank mRNA | AK034454.1 | - | 2 | 1863 | ||
| AK034584.1 | - | 2 | 1886 | |||
| AK046410.1 | - | 2 | 1846 | |||
| AK052931.1 | - | 2 | 1843 | |||
| BC117813.2 | - | 2 | 1082 | 40092671 | ||

Arsj
Genomic Map