siRNA Details
| Gene Symbol: Tdrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173744.4 | NP_776105.1 | 3 | 1089 | ||
| GenBank mRNA | AK011462.1 | - | 1 | 621 | ||
| AK077810.1 | - | 3 | 1003 | |||
| AK166875.1 | - | 3 | 1000 | |||
| AK166972.1 | - | 3 | 996 | |||
| BC099613.1 | - | 3 | 994 | 30031671 | ||

Tdrp
Genomic Map