siRNA Details
| Gene Symbol: Ilvbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173751.4 | NP_776112.1 | 12 | 1492 | ||
| GenBank mRNA | AK087956.1 | - | 11 | 1366 | ||
| AK146075.1 | - | 11 | 1420 | |||
| AK169899.1 | - | 11 | 1418 | |||
| AK218076.1 | - | 1 | 42 | |||
| BC052054.1 | - | 11 | 1826 | |||
| BC056459.1 | - | 12 | 1450 | 5718157 | ||

Ilvbl
Genomic Map